Learn More
IDEX Health & Science Upchurch 4 WAY PEEK BULKHEAD FLOW VALVE
Catalog No. NC2045372
Encompass_Preferred
4 Way PEEK Bulkhead Flow Valve1/4-28PEEK/PTFE(psi) 500(bar) 34(in) 1/16
Catalog No. NC2045372 Supplier IDEX Health & Science Upchurch Supplier No. V101T
4 Way PEEK Bulkhead Flow Valve1/4-28PEEK/PTFE(psi) 500(bar) 34(in) 1/16