Learn More
COLE PARMER INSTRUMENT COMPANY Idex V-101D Manual Switching Valve, Double Diagonal, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Catalog No. NC1760459
Encompass
Easily switch between two fluid paths without disconnecting the lines. Enable connections among multiple fluid systems. Double diagonal, bulkhead, and swept volume of 12.4 uL. 1/4-28 flat-bottom ports for 1/16" OD tubing.
Catalog No. NC1760459 Supplier COLE PARMER INSTRUMENT COMPANY Supplier No. 0201488
Easily switch between two fluid paths without disconnecting the lines. Enable connections among multiple fluid systems. Double diagonal, bulkhead, and swept volume of 12.4 uL. 1/4-28 flat-bottom ports for 1/16" OD tubing.