Learn More
Analytical Sales & Services 4-Way Rack
Catalog No. 5000805539
Encompass
4-Way Rack Tubes: 50mL, 15mL, 0.5mL, 1.5/2.0mL Wells: 4, 12, 32, 32 Well Diameter: 30mm, 17.5mm, 8.3mm, 11.6mm Assorted Colors
Catalog No. 50-008-05539 Supplier Analytical Sales & Services Supplier No. 4WRACK
4-Way Rack Tubes: 50mL, 15mL, 0.5mL, 1.5/2.0mL Wells: 4, 12, 32, 32 Well Diameter: 30mm, 17.5mm, 8.3mm, 11.6mm Assorted Colors