Learn More
Andwin Scientific 4-WAY RACK HOLDS TUBE 0.5-50ML
SDP
Catalog No. 50819915
Encompass_Preferred
With a special design of being able to stand in four different sides, the neon green 4-way rack can hold and support different sizes of tubes. The rack can be used with four 50 ml tubes, twelve 15 ml tubes, thirty-two 1.5 or 2.0 ml tubes, or thirty-two 0.5 ml tubes. The racks can also be joined with another rack in order to support several sets of 50 and 15 ml tubes.
Catalog No. 50-819-915 Supplier Andwin Scientific Supplier No. 23048042
With a special design of being able to stand in four different sides, the neon green 4-way rack can hold and support different sizes of tubes. The rack can be used with four 50 ml tubes, twelve 15 ml tubes, thirty-two 1.5 or 2.0 ml tubes, or thirty-two 0.5 ml tubes. The racks can also be joined with another rack in order to support several sets of 50 and 15 ml tubes.