Learn More
Andwin Scientific 4-WAY RACK HOLDS TUBES 0.5 TO
SDP
Catalog No. 50819914
Encompass_Preferred
With a special design of being able to stand in four different sides, the neon blue 4-way rack can hold and support different sizes of tubes. The rack can be used with four 50 ml tubes, twelve 15 ml tubes, thirty-two 1.5 or 2.0 ml tubes, or thirty-two 0.5 ml tubes. The racks can also be joined with another rack in order to support several sets of 50 and 15 ml tubes.
Catalog No. 50-819-914 Supplier Andwin Scientific Supplier No. 23048041
With a special design of being able to stand in four different sides, the neon blue 4-way rack can hold and support different sizes of tubes. The rack can be used with four 50 ml tubes, twelve 15 ml tubes, thirty-two 1.5 or 2.0 ml tubes, or thirty-two 0.5 ml tubes. The racks can also be joined with another rack in order to support several sets of 50 and 15 ml tubes.