Learn More
Alkali Scientific 4-Way Tube Rack (4x50mL, 12x15mL, 32x1.5mL/0.5mL), Rainbow Pack, 5/cs
SDP
Catalog No. 502335759
Encompass_Preferred
ASI's bench-top tube racks are made of 100% polypropylene. FR5015 is a 4-way rack that can be used to hold 4 x 50mL, 12 x 15mL or 32 x Microtubes (0.5, 1.5 or 2.0mL). They can be snap-linked together to form as large a rack as is needed. Available in a "Rainbow Pack" of five colors.
Catalog No. 50-233-5759 Supplier Alkali Scientific Supplier No. FR5015
ASI's bench-top tube racks are made of 100% polypropylene. FR5015 is a 4-way rack that can be used to hold 4 x 50mL, 12 x 15mL or 32 x Microtubes (0.5, 1.5 or 2.0mL). They can be snap-linked together to form as large a rack as is needed. Available in a "Rainbow Pack" of five colors.