Learn More
Research Products International Corp Flipper 4-Way Tube Racks, Natural, 3 1/2 x 2 x 7 Inches, 5 per Case
SDP
Catalog No. 50476488
Encompass
Flipper 4-Way Tube Racks are designed to hold a variety of tube sizes and styles.
Racks can be rotated to any of four positions to accommodate:
  • 4-50 ml conical tubes
  • 32-12x75 or 1.5 ml tubes
  • 12-15 ml conical tubes
  • 32-0.5 ml micro-tubes
Autoclavable racks are constructed of durable polypropylene and can be used in water baths and freezers.
Catalog No. 50-476-488 Supplier Research Products International Corp Supplier No. 167000
Flipper 4-Way Tube Racks are designed to hold a variety of tube sizes and styles.
Racks can be rotated to any of four positions to accommodate:
  • 4-50 ml conical tubes
  • 32-12x75 or 1.5 ml tubes
  • 12-15 ml conical tubes
  • 32-0.5 ml micro-tubes
Autoclavable racks are constructed of durable polypropylene and can be used in water baths and freezers.