Learn More
MatTek Corporation (Case of 12) Chambered Cell Culture Slides, 4-well
Supplier:  MatTek Corporation CCS4
Encompass
4-well Chambered Cell Culture Slide, Quantity 12With a proven track record of quality and convenience, MatTek dishes provide the optics of glass in standard size microscopy dishes and plates. Cited in thousands of scientific publications, MatTek dishes are a long-standing favorite of lab teams worldwide for their availability, ease-of-use, and the brilliant images they produce.These Cell Chambered Glass Slides from MatTek Life Sciences offer the unique benefit of culturing your cells in multiple different conditions while minimizing other variables on a single standard glass slide. The easy “snap-on, snap-off” chamber slide uses no adhesive, and does not require a special tool for removal. The naturally charged surface is great for adherent cells. This lends itself to a convenient method for your microscopic observations.Compare to Ibidi , Cellvis , Chemglass , or Nunc products.
Catalog No. NC1844411