Learn More
CELLTREAT Scientific Products 4 Well Non-treated Plate, Sterile
SDP
Catalog No. 501149115
Encompass_Preferred
CELLTREAT Scientific Products 4 Well Non-Treated Plate with Lid. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. 4 well plate is a smaller footprint option which offers well size similar to that of a 24 well plate, but has expanded well spacing to reduce cross contamination. Has a working volume of 500-800¿L. RNase/DNase Free and non pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 50/case.
Catalog No. 50-114-9115 Supplier CELLTREAT Scientific Products Supplier No. 229503
Add to Cart
Add to Cart
CELLTREAT Scientific Products 4 Well Non-Treated Plate with Lid. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. 4 well plate is a smaller footprint option which offers well size similar to that of a 24 well plate, but has expanded well spacing to reduce cross contamination. Has a working volume of 500-800¿L. RNase/DNase Free and non pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 50/case.