Learn More
Electron Microscopy Sciences NUNC™ 4 Well Slide Processing Dish
SDP
Catalog No. 5031452
Encompass_Preferred
The 4-well slide processing dish holds four 25 x 75 mm slides. It is sterile and has a clear polysturene dish and lid.
Catalog No. 50-314-52 Supplier Electron Microscopy Sciences Supplier No. 63496
The 4-well slide processing dish holds four 25 x 75 mm slides. It is sterile and has a clear polysturene dish and lid.