Learn More
Air Science Usa Ductless Fume Hood, 48" / 1200mm Nominal Width, 115V 60Hz
SDP
Catalog No. NC1583179
Encompass_Preferred
Purair Basic Ductless Fume Hood (P5-48-XT), 48" / 1200mm Nominal Width, Standard Depth, 115V 60Hz (other voltages available). The Purair Basic ductless fume hoods are designed to provide high level performance features at an affordable price. Purair Basic ductless hoods are found in research laboratories, educational facilities, and forensic laboratories throughout the world and are designed to protect the user and the environment from hazardous vapors generated on the work surface. At the heart of the Purair fume hoods product line is the innovative Multiplex Filtration Technology that creates a safe work environment over the widest range of applications in the industry. Units Requires qty. 2 Asts-Xxx Carbon and/or HEPA filters sold separately. The Purair Basic can be equipped with a single activated carbon main filter or with a stacked configuration which combines two main filters, each to adsorb one or more specific vapors, family of vapors, or particulates
Catalog No. NC1583179 Supplier Air Science Usa Supplier No. P548XT
Purair Basic Ductless Fume Hood (P5-48-XT), 48" / 1200mm Nominal Width, Standard Depth, 115V 60Hz (other voltages available). The Purair Basic ductless fume hoods are designed to provide high level performance features at an affordable price. Purair Basic ductless hoods are found in research laboratories, educational facilities, and forensic laboratories throughout the world and are designed to protect the user and the environment from hazardous vapors generated on the work surface. At the heart of the Purair fume hoods product line is the innovative Multiplex Filtration Technology that creates a safe work environment over the widest range of applications in the industry. Units Requires qty. 2 Asts-Xxx Carbon and/or HEPA filters sold separately. The Purair Basic can be equipped with a single activated carbon main filter or with a stacked configuration which combines two main filters, each to adsorb one or more specific vapors, family of vapors, or particulates