Learn More
Chemglass Life Sciences 4in Diameter X 1/8in Thick Borosilicate Disc
SDP
Catalog No. CG190616
Encompass_Preferred
Plate glass, disc, borosilicate, 4in Diameter X 1/8in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.
Catalog No. CG190616 Supplier Chemglass Life Sciences Supplier No. CG190616
Plate glass, disc, borosilicate, 4in Diameter X 1/8in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.