Learn More
Uline 4 x 6 3 Mil Slider Zip Bags,100
Catalog No. NC1570209
Encompass_Preferred
Slide lock glides easily across bag protecting contents from dirt and moisture. Durable and reusable. Easy open, easy close. Bags are pre-opened for quick handling. 100% virgin polyethylene film. FDA and USDA compliant for food contact.
Catalog No. NC1570209 Supplier Uline Supplier No. S10832
Slide lock glides easily across bag protecting contents from dirt and moisture. Durable and reusable. Easy open, easy close. Bags are pre-opened for quick handling. 100% virgin polyethylene film. FDA and USDA compliant for food contact.