Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 4mm ID X 6mm OD
SDP
Catalog No. CGQ031003
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 4mm ID X 6mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031003 Supplier Chemglass Life Sciences Supplier No. CGQ031003
Graded Seal, Quartz to Borosilicate, 4mm ID X 6mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.