Learn More
Cayman Chemical 4 Z 10 Z 13 Z 16 ZDocosatetra
SDP
Catalog No. 502889967
Encompass_Preferred
A polyunsaturated fatty acid methyl acid; a minor fatty acid component of rat testis lipids.
Catalog No. 50-288-9967 Supplier Cayman Chemical Supplier No. 100068665 MG
Add to Cart
Add to Cart
A polyunsaturated fatty acid methyl acid; a minor fatty acid component of rat testis lipids.