Learn More
Medchemexpress LLC 4-[(Z)-[4-oxo-2-sulfanylidene-3-[[3-(trifluoromethyl)phenyl]methyl]-1,3-thiazolidin-5-ylidene]methyl]benzoic acid | 1073612-91-5 | MFCD31619349 | 99.6% | 423.43 g/mol | C19H12F3NO3S2 | 100 MG
SDP
Catalog No. 5000244900
Encompass_Preferred
CY-09 is a selective, direct inhibitor of the NLRP3 inflammasome used for biochemical and cellular research. It binds the ATP-binding motif of the NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, preventing inflammasome assembly and activation. For research use only.
  • Selective NLRP3 inhibitor targeting the NACHT domain
  • Inhibits NLRP3 ATPase activity to block inflammasome activation
  • High purity compound suitable for biochemical and cellular assays
  • Available as a solid powder, stable under recommended storage conditions
  • Supplied in research-scale pack sizes for laboratory experiments
Catalog No. 50-002-44900 Supplier Medchemexpress LLC Supplier No. HY103666100MG
CY-09 is a selective, direct inhibitor of the NLRP3 inflammasome used for biochemical and cellular research. It binds the ATP-binding motif of the NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, preventing inflammasome assembly and activation. For research use only.
  • Selective NLRP3 inhibitor targeting the NACHT domain
  • Inhibits NLRP3 ATPase activity to block inflammasome activation
  • High purity compound suitable for biochemical and cellular assays
  • Available as a solid powder, stable under recommended storage conditions
  • Supplied in research-scale pack sizes for laboratory experiments