Learn More
Medchemexpress LLC 4-((Z)-4-oxo-2-thioxo-3-(3-(trifluoromethyl)benzyl)thiazolidin-5-ylidene)methyl benzoic acid | 1073612-91-5 | MFCD31619349 | 99.6% | 423.43 g/mol | C19H12F3NO3S2 | 50 MG
SDP
Catalog No. 5000246723
Encompass_Preferred
CY-09 is a selective, direct small-molecule inhibitor of the NLRP3 inflammasome that binds the ATP-binding motif of the NLRP3 NACHT domain, inhibiting NLRP3 ATPase activity and suppressing inflammasome assembly and activation. It is supplied as a solid for research use in studies of inflammatory signaling and innate immunity.
  • Selective NLRP3 inhibitor that binds the NLRP3 ATP-binding motif.
  • Inhibits NLRP3 ATPase activity to block inflammasome assembly.
  • Applicable to research on inflammation, innate immunity, and disease models.
  • Solid, light yellow to yellow appearance; high purity suitable for biochemical assays.
  • Recommended storage: powder at -20°C for long term stability.
Catalog No. 50-002-46723 Supplier Medchemexpress LLC Supplier No. HY10366650MG
CY-09 is a selective, direct small-molecule inhibitor of the NLRP3 inflammasome that binds the ATP-binding motif of the NLRP3 NACHT domain, inhibiting NLRP3 ATPase activity and suppressing inflammasome assembly and activation. It is supplied as a solid for research use in studies of inflammatory signaling and innate immunity.
  • Selective NLRP3 inhibitor that binds the NLRP3 ATP-binding motif.
  • Inhibits NLRP3 ATPase activity to block inflammasome assembly.
  • Applicable to research on inflammation, innate immunity, and disease models.
  • Solid, light yellow to yellow appearance; high purity suitable for biochemical assays.
  • Recommended storage: powder at -20°C for long term stability.