Learn More
Cayman Chemical 4 Z 7 Z 10 Z 13 Z 16 Z-N 100ug
SDP
Supplier:  Cayman Chemical 14893100 UG
Encompass_Preferred
An internal standard for the quantification of 4(Z),7(Z),10(Z),13(Z),16(Z)-nonadecapentaenoic acid by GC- or LC-MS
Catalog No. 50-296-9395