Learn More
Cayman Chemical 4 Z 7 Z 10 Z 13 ZHexadec 100UG
SDP
Supplier:  Cayman Chemical 9002202100 UG
Encompass_Preferred
A PUFA; inhibits the development of starfish embryos past the one-cell stage at 25 UG/ml
Catalog No. 50-289-5544