Learn More
Cayman Chemical 4 Z 7 Z 10 ZTrIDcatrIeno 100ug
SDP
Catalog No. 502970042
Encompass_Preferred
An internal standard for the quantification of 4(Z),7(Z),10(Z)-tridecatrienoic acid by GC- or LC-MS
Catalog No. 50-297-0042 Supplier Cayman Chemical Supplier No. 14895100 UG
An internal standard for the quantification of 4(Z),7(Z),10(Z)-tridecatrienoic acid by GC- or LC-MS