Learn More
Cayman Chemical 4 Z 7 ZDecadIenoIc ACDd5 100ug
SDP
Catalog No. 502969515
Encompass_Preferred
An internal standard for the quantification of 4(Z),7(Z)-decadienoic acid by GC- or LC-MS
Catalog No. 50-296-9515 Supplier Cayman Chemical Supplier No. 14896100 UG
An internal standard for the quantification of 4(Z),7(Z)-decadienoic acid by GC- or LC-MS