Learn More
Ace Glass, Inc. 40/25 socket joint, stainless steel, unthreaded
SDP
Supplier:  Ace Glass, Inc. 765858
Encompass
40/25, unthreaded, socket joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-975-301