Learn More
Ace Glass, Inc. Joint clips, 40/35, Keck; type, POM acetal resin, not for positive pressure use, dark yellow, pk/10
SDP
Catalog No. 50979348
Encompass
Catalog No. 50-979-348 Supplier Ace Glass, Inc. Supplier No. 759840