Learn More
Uline 40-45 GAL 3MIL 40X46 BIOHAZ BAG 75/case
Catalog No. NC0932388
Encompass
BIOHAZARD BAGS LINEAR LOW DENSITY Isolate hazardous items in hospitals, doctor's offices and healthcare facilities. Puncture resistant. Flat seal bottom. Flat packed in carton.40 x 46 in. 40-45 Gallon 3.0mil thick 75 per carton
Catalog No. NC0932388 Supplier Uline Supplier No. S17202
BIOHAZARD BAGS LINEAR LOW DENSITY Isolate hazardous items in hospitals, doctor's offices and healthcare facilities. Puncture resistant. Flat seal bottom. Flat packed in carton.40 x 46 in. 40-45 Gallon 3.0mil thick 75 per carton