Learn More
Chemglass Life Sciences 40/50 Outer Joint, Full Length, on 45mm OD x 2mm W tubing
SDP
Catalog No. CG10109
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 40/50 Outer Joint, Full Length, on 45mm OD x 2mm W tubing.
Catalog No. CG10109 Supplier Chemglass Life Sciences Supplier No. CG10109
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 40/50 Outer Joint, Full Length, on 45mm OD x 2mm W tubing.