Learn More
Bio Rad Laboratories 40% BIS ACRYLAMIDE SOLUTION
Supplier:  Bio Rad Laboratories 1610148
Encompass
40% Acrylamide/Bis Solution, 37.5:1, 500mL, Ready to use, Made from 99.9% pure acrylamide and bis-acrylamide
Catalog No. NC9427788