Learn More
National Safety Apparel™ 40 Cal ArcGuard™ Compliance 49 in. Arc Flash Coat
SDP
Catalog No. 17001260 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-260 5X-Large
17-001-253 Small
17-001-254 Medium
17-001-255 Large
17-001-256 X-Large
17-001-257 2X-Large
17-001-258 3X-Large
17-001-259 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-260 Supplier National Safety Apparel™ Supplier No. C04UQUQ405X49
Technology advanced Westex™ UltraSoft offers advanced flame resistant fabric and provides comfort, protection, and durability.
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 9 oz. Westex™ UltraSoft™ Sateen, Double Layer
  • Breathable FR fabric will not melt, drip, or ignite
  • Generous cut allows for unrestrictive movement
  • FR hook and loop closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Available in a convenient kit
  • Made in the USA
Specifications
Closure Type FR Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size 5X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269