Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ 40 Cal ArcGuard™ Compliance Arc Flash Leggings
SDP

Catalog No. 170012052 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2052 2X-Large
17-001-2050 Large
17-001-2051 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-001-2052 Supplier National Safety Apparel™ Supplier No. L05UQUQ402X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compliance leggings with arc rating of 40 cal/cm2

  • 9 oz. Double Layer UltraSoft™
  • Adjustable FR hook and loop closure
  • For use with 40 cal FR long coat

Specifications

Size 2X-Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Ankle Style Open
Seam Type Stitched
Disposable Reusable
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.