Learn More
National Safety Apparel™ 40 Cal ArcGuard™ Compliance Bib Overalls
SDP
Catalog No. 17001356 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-356 5X-Large
17-001-349 Small
17-001-350 Medium
17-001-351 Large
17-001-352 X-Large
17-001-353 2X-Large
17-001-354 3X-Large
17-001-355 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-356 Supplier National Safety Apparel™ Supplier No. C45UQUQ405X32
Bib overalls with quick disconnect and easy adjustable shoulder straps for quick entry
  • Arc Rating: 40 cal/cm²
  • CAT 4 Fabric/Material: 9 oz. Westex™ UltraSoft™ Sateen, Double Layer
  • Breathable FR fabric
  • FR fabric will not melt, drip, or ignite
  • Quick disconnect and easy adjustable shoulder straps for quick entry
  • Generous cut allows for unrestricted movement
  • Quick disconnect buckles at each shoulder strap
  • Inherently flame resistant for FR protection
  • Garments embroidered with arc rating
  • Available in a convenient kit
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Closure Type Buckle
Color Navy
Material Cotton, Nylon
Seam Type Stitched
Size 5X-Large
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Disposable Reusable