Learn More
National Safety Apparel™ 40 Cal ArcGuard™ Compliance CrossVent Hood with PureView Faceshield
SDP
Catalog No. 17001736 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-736 Color: Navy
1 options
Catalog No. 17-001-736 Supplier National Safety Apparel™ Supplier No. H65UQUQFAN40PV
CrossVent hood with pureview faceshield
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 9 oz. Westex™ UltraSoft™ Sateen, Double Layer
  • Breathable FR fabric
  • FR fabric will not melt, drip, or ignite
  • Anti-fog faceshield
  • Fans are attached with FR hook and loop
  • Fans are powered by 9-volt batteries
  • Hard hat included with slotted adapter
  • Hood fans provide a gentle and quiet breeze that keeps you cool and decreases lens fogging
  • Features PureView™ single-layer, highly transparent faceshield technology
  • Made in the USA
Specifications
Size Medium
Material Cotton, Nylon
Color Navy
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269