Learn More
National Safety Apparel™ 40 Cal ArcGuard™ Compliance Hood with PureView Faceshield
SDP
Catalog No. 17001733 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-733 Color: Navy
1 options
Catalog No. 17-001-733 Supplier National Safety Apparel™ Supplier No. H65UQUQ40HATPV
Made of a breathable, FR Fabric that won't melt, drip or ignore. The arc flash protection hood includes an anti-fog and scratch resistant PureView™ face shield for enhanced visibility and safety, as well as a hard hat with a slotted adapter
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 9 oz. Westex™ UltraSoft™ Sateen, Double Layer
  • Breathable FR fabric
  • FR fabric will not melt, drip, or ignite
  • Premium anti-fog and scratch resistant coating on faceshield
  • Hard hat with slotted adapter included
  • PureView™ faceshield technology which features a single layer lens and greater range of visibility
  • Made in the USA
Specifications
Size Medium
Material Cotton, Nylon
Color Navy
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269, ANSI/ISEA Z87.1 (Impact Rating Z81+)