Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Arc Flash Coat
SDP
Catalog No. 17001213 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-213 Small
17-001-214 Medium
17-001-215 Large
17-001-216 X-Large
17-001-217 2X-Large
17-001-218 3X-Large
17-001-219 4X-Large
17-001-220 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-213 Supplier National Safety Apparel™ Supplier No. C04NPQHLTDSM32
Add to Cart
Add to Cart
Arc flash coats are equipped with hook and loop front closure with drawstring waist.
  • 50% lighter than standard 40 cal/cm2 arc flash kits
  • Stand up collar for extra coverage, can be turned down
  • Nomex knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • FR fabric will not melt, drip, or ignite
  • Available in a convenient kit
Specifications
Closure Type FR Hook and Loop with Buckle
Color Olive Green
Disposable Reusable
Gender Unisex
Material RevoLite™
Neck Style Collared
Size Small
Resistance Flame
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269