Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite Arc Flash Hood with PureView Faceshield
SDP
Catalog No. 17001731 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Olive green
Catalog No.
17-001-731 Color: Olive green
1 options
Catalog No. 17-001-731 Supplier National Safety Apparel™ Supplier No. H65NPQHHATPV
The features in the 40 cal arc flash hood make for a clearer view: Comfortable and Lightweight protection – RevoLite™ breathable flame resistant and arc rated fabric is 50% lighter than standard 40 cal fabric options
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal/cm²
  • Arc Flash PPE
  • Inherently flame resistant
  • Premium anti-fog & scratch resistant coating on faceshield
  • FR fabric will not melt, drip, or ignite
  • PureView™ faceshield technology features a greater range of visibility
  • Made in the USA
Specifications
Size Medium
Material RevoLite™
Color Olive Green
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269