Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Arc Flash Kit with Lift Front Hood
SDP

Catalog No. 17355206 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size: Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-355-206 Size: Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-355-206 Supplier National Safety Apparel™ Supplier No. KIT4SCLT40NGLF_ _
This item is not returnable. View return policy

National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Arc Flash Kit with Lift Front Hood is designed to be 50% lighter than a standard 40 cal kits.

  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots

Specifications

Arc Flash Rating 40 cal/cm²
Gender Unisex
Resistance Flame
For Use With (Application) Arc Flash
Neck Style Hooded
Color Green
Certifications/Compliance NFPA 70E/ASTM F1506/CSA Z462 for Arc Flash, OSHA 1910.269
Closure Type Snap
Material RevoLite™
Seam Type Serged
Size Universal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.