Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Bib Overalls
SDP
Catalog No. 17001325 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-325 Small
17-001-326 Medium
17-001-327 Large
17-001-328 X-Large
17-001-329 2X-Large
17-001-330 3X-Large
17-001-331 4X-Large
17-001-332 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-325 Supplier National Safety Apparel™ Supplier No. C45NPQHLTSM32
Add to Cart
Add to Cart
RevoLite; bib overalls are 50% lighter than standard 40 cal/cm2 Arc Flash PPE
  • Inherently flame resistant for FR protection
  • FR fabric will not melt, drip, or ignite
  • Quick disconnect and easy adjustable shoulder straps
  • Heavy duty 26 in. zipper closure on each leg
  • Expandable leg opening to fit over boots
  • Garments embroidered with arc rating
  • Available in a convenient kit
Specifications
Closure Type FR Hook and Loop with Buckle
Color Olive
Material RevoLite™
Seam Type Stitched
Size Small
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 40 cal/cm²
Product Type Bib Overalls
Waist Style Standard
Resistance Flame
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Disposable Reusable