Learn More
Perkin Elmer US LLC 40 mL Headspace Convenience Kit - Amber Glass Vial with Open Top Grey Polypropylene Screw Cap and PTFE/Silicone Septa, 72/pk
Catalog No. 502104270
Encompass_Preferred
Vial Material: Cap Type Cap Material Septa Material Quantity Amber Glass Screw Top, Open Grey Polypropylene PTFE/Silicone 72
Catalog No. 50-210-4270 Supplier Perkin Elmer US LLC Supplier No. N6352030
Vial Material: Cap Type Cap Material Septa Material Quantity Amber Glass Screw Top, Open Grey Polypropylene PTFE/Silicone 72