Learn More
Nanocomposix BioReady Gold Nanospheres - Bare (Citrate) - 40 nm, 20 OD in aq. 0.02 mM sodium citrate, 5 mL
SDP
Catalog No. NC1564785
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.40 nm spherical gold nanoparticles for passive adsorption combine strong plasmonic optical properties with surface chemistry designed for passive adsorption. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC1564785 Supplier Nanocomposix Supplier No. AUCR405M
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.40 nm spherical gold nanoparticles for passive adsorption combine strong plasmonic optical properties with surface chemistry designed for passive adsorption. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.