Learn More
Chemglass Life Sciences O-Ring Joint, 40mm ID, 45mm Tube OD , O-Ring Size: 226, Clamp Size: 65
SDP
Catalog No. CG12408
Encompass_Preferred
O-Ring Joint, 40mm ID, 45mm Tube OD , O-Ring Size: 226, Clamp Size: 65. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.
Catalog No. CG12408 Supplier Chemglass Life Sciences Supplier No. CG12408
O-Ring Joint, 40mm ID, 45mm Tube OD , O-Ring Size: 226, Clamp Size: 65. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.