Learn More
Electron Microscopy Sciences 40 Place Rectangular Mailing Rack for 0.5 ml Tubes, 4½" x 2⅜" x 1-5/16"
SDP
Catalog No. 50949555
Encompass_Preferred
Designed to be used in water/ice baths complete with rust-proof anodized aluminum rack. Removable cover keeps tubes closed while boiling. Handle for easy removal from water baths. Will not rust. 40-place rack holds 0.5ml tubes.
Catalog No. 50-949-555 Supplier Electron Microscopy Sciences Supplier No. 7238024
Designed to be used in water/ice baths complete with rust-proof anodized aluminum rack. Removable cover keeps tubes closed while boiling. Handle for easy removal from water baths. Will not rust. 40-place rack holds 0.5ml tubes.