Learn More
CEM Corporation 40-PLACE STARTER SET 75ML
SDP
Catalog No. NC1971023
Encompass
  • Engineered for high-throughput applications
  • Includes 75 mL PFA vessels
  • Suitable for microwave digestion and solvent extraction
  • Compatible with iWave and IR control
  • User-friendly assembly
  • Designed for moderate temperature and pressure conditions
  • Ideal for sample matrices including plants, foods, vitamins, and environmental samples
  • Cost-effective solution for routine sample preparation
  • High throughput capabilities
Catalog No. NC1971023 Supplier CEM Corporation Supplier No. 907397
Add to Cart
Add to Cart
Request a Quote
  • Engineered for high-throughput applications
  • Includes 75 mL PFA vessels
  • Suitable for microwave digestion and solvent extraction
  • Compatible with iWave and IR control
  • User-friendly assembly
  • Designed for moderate temperature and pressure conditions
  • Ideal for sample matrices including plants, foods, vitamins, and environmental samples
  • Cost-effective solution for routine sample preparation
  • High throughput capabilities