Learn More
Agilent Technologies 40 RXN PFUULTRA II FUSION HS
Catalog No. NC9387714
Encompass
PfuUltra II Fusion HS DNA Polymerase; 40 rxn; 1mL; Store at -20 deg.C; Extended length capability up to 19 kb
Catalog No. NC9387714 Supplier Agilent Technologies Supplier No. 600670
PfuUltra II Fusion HS DNA Polymerase; 40 rxn; 1mL; Store at -20 deg.C; Extended length capability up to 19 kb