Learn More
Indiana Fluid Systems Technologies Brass 1-Piece 40 Series 3-Way Ball Valve
Catalog No. NC1828692
Encompass
Brass 1-Piece 40 Series 3-Way Ball Valve, 0.15 Cv, 1/8 in. Swagelok Tube Fitting.
Catalog No. NC1828692 Supplier Indiana Fluid Systems Technologies Supplier No. B41XS2
Brass 1-Piece 40 Series 3-Way Ball Valve, 0.15 Cv, 1/8 in. Swagelok Tube Fitting.