Learn More
Hampton Research Additive Screen 90 (H6) 1.0ml
SDP
Catalog No. 500923
Encompass
40% v/v 1-Propanol in the amount of 1ml. It is formulated in Type 1+ ultrapure water: 18.2 megaohm-cm resistivity at 25°C, < 5 ppb Total Organic Carbon, bacteria free (<1 Bacteria (CFU/ml)), pyrogen free (<0.03 Endotoxin (EU/ml)), RNase-free (< 0.01 ng/mL) and DNase-free (< 4 pg/µL).
Catalog No. 50-092-3 Supplier Hampton Research Supplier No. HR242890
40% v/v 1-Propanol in the amount of 1ml. It is formulated in Type 1+ ultrapure water: 18.2 megaohm-cm resistivity at 25°C, < 5 ppb Total Organic Carbon, bacteria free (<1 Bacteria (CFU/ml)), pyrogen free (<0.03 Endotoxin (EU/ml)), RNase-free (< 0.01 ng/mL) and DNase-free (< 4 pg/µL).