Learn More
Teknova 40% (v/v) Glycerol. 100mL, Sterile
SDP
Catalog No. 502387862
Encompass
Glycerol solution is a versatile solvent that can be used in a variety of applications including electrophoresis, bacterial stock cultures, as an additive as a carbon source, and stabilizer.
Catalog No. 50-238-7862 Supplier Teknova Supplier No. G1801
Glycerol solution is a versatile solvent that can be used in a variety of applications including electrophoresis, bacterial stock cultures, as an additive as a carbon source, and stabilizer.