Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Glass Autosampler Vial Inserts

Catalog No. 13622218 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-622-218 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-622-218
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fisherbrand™ glass autosampler vial inserts are available in precision point and flat bottom profiles and are compatible with 9 mm HPLC and GC autosampler vials.

Nominal O.D. of 6 mm to fit Target wide-opening vials.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Diameter (Metric) Outer 6 mm
Material Glass
Volume (Metric) 400 μL
No. per Pack 500
Style Flat Bottom
Volume (Metric) Residual < 25 μL
Product Line Fisherbrand
Volume (Metric) Total 500 μL
Volume (Metric) Usable 450 μL
Height (Metric) 31 mm
Product Type Autosampler Vial Insert
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.