Learn More
Accuflow Systems Altafluor 400 PFA - available in coiled hose and convoluted tubing constructions - 400 PFA 1/2" X 5/8"- 100 FT
Catalog No. 501355172
Encompass_Preferred
Altafluor 400 PFA (Perfluoroalkoxy) is the product of choice for applications involveing extreme chemical resistance combined with high temperature. Altifluor 400 is made exclusively from 100% virgin grade high performance resin used to resist stress cracking. The highest molecular weight resins available giving it superior physical properties making it suitable for even the most demanding applications. Chemically inert to nearly all industrial chemicals and solvents. It has a higher thermal stability than FEP. is translucent in appearance. It is FDA compliant for food contact, FDA 21 CFR 177.1550. It meets the requirements of USP Class VI, is non-flammable and has a moisture absorption of nearly zero. Altafluor is compatible with flare of conventional fittings.
Catalog No. 50-135-5172 Supplier Accuflow Systems Supplier No. 4000625062100
Altafluor 400 PFA (Perfluoroalkoxy) is the product of choice for applications involveing extreme chemical resistance combined with high temperature. Altifluor 400 is made exclusively from 100% virgin grade high performance resin used to resist stress cracking. The highest molecular weight resins available giving it superior physical properties making it suitable for even the most demanding applications. Chemically inert to nearly all industrial chemicals and solvents. It has a higher thermal stability than FEP. is translucent in appearance. It is FDA compliant for food contact, FDA 21 CFR 177.1550. It meets the requirements of USP Class VI, is non-flammable and has a moisture absorption of nearly zero. Altafluor is compatible with flare of conventional fittings.