Learn More
Tubewriter / California Advanced Labeling, Inc TubeWriter 400 - Standard Rack for Printing Labels Directly on the Side of 1.5-2.0mL Microcentrifuge Tubes
SDP
Catalog No. NC2956947 Shop All California Advanced Labeling TubeWriter Products
Encompass
1.5-2.0mL Microcentrifuge Tube printing racks ensure correct alignment and hold securely while printing labels directly on the side of the tube. The TubeWriter 400 racks are designed for rapid switching between labware types.
Catalog No. NC2956947 Supplier Tubewriter / California Advanced Labeling, Inc Supplier No. PS1500MCEL400
1.5-2.0mL Microcentrifuge Tube printing racks ensure correct alignment and hold securely while printing labels directly on the side of the tube. The TubeWriter 400 racks are designed for rapid switching between labware types.