Learn More
Sino Biological Influenza A H5N1 (A/Duck/Hong Kong/p46/97) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40001V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met1-Glu348) of the influenza hemagglutinin (A/Duck/Hong Kong/p46/97 (H5N1)) (AAF02306.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0061