Learn More
Sino Biological Influenza A H9N2 (A/duck/Hong Kong/448/78) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40003V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Hong Kong/448/78(H9N2)) hemagglutinin (AAO46079.1) (Met1-Lys523), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0064