Learn More
Sino Biological Influenza A H5N1 (A/Xinjiang/1/2006) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40004V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza hemagglutinin (A/Xinjiang/1/2006 (H5N1)) (ACJ68614.1) (HA1+HA2, uncleaved), cleavage site mutated (RRRKKR-TETR) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0068